Learn More
Sino Biological EGFR / HER1 / ErbB1 Antibody, Rabbit PAb DFS Item
Manufacturer:  Sino Biological 10001RP01400
Produced in rabbits immunized with purified, recombinant Human EGFR / ErbB / HER1 (rh EGFR / ErbB / HER1; Catalog#10001-H08H; Met 1-Ser 645; NP_005219). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-161-0283