Learn More
Sino Biological EGFR Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 10001T48200
Produced in rabbits immunized with purified, recombinant Human EGFR (rh EGFR; Catalog#10001-H08B; NP_005219; Met1-Lys642). EGFR specific IgG was purified by Human EGFR affinity chromatography.
Catalog No. 50-161-0291