Learn More
Sino Biological Anti-HA Tag Antibody, Mouse MAb DFS Item
Manufacturer:  Sino Biological 100028MM105
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, a synthetic peptide corresponding to the HA-tag sequence. The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-161-0295