Learn More
Sino Biological Influenza A H5N1 (A/VietNam/1203/2004) Hemagglutinin Protein (HA1 Subunit) (His & Fc Tag) DFS Item
Manufacturer:  Sino Biological 10003V06H150
A DNA sequence encoding an N-terminal segment (Met 1-Arg 342) of Influenza A Virus H5N1 Hemagglutinin (AAW80717.1),termed as HA1, was fused with the C-terminal His-tagged Fc region of mouse IgG1 at the C-terminus.
Catalog No. 50-161-3734
Due to product restrictions, please to purchase or view availability for this product.