Learn More
Sino Biological Influenza A H5N1 (A/VietNam/1203/2004) Hemagglutinin / HA Protein (His & Fc Tag) DFS Item
Manufacturer:  Sino Biological 10003V06H310
A DNA sequence encoding the extracellular domain of hemagglutinin (Influenza A virus (A/Viet Nam/1203/2004 (H5N1)) (AAW80717.1) (Met 1-Gln 531) with cleavage site mutated (RERRRKKR→TETR,HA1+HA2,uncleaved) was fused with the C-terminal polyhistidine-tagged Fc region of mouse IgG1 at the C-terminus.
Catalog No. 50-161-3735
Due to product restrictions, please to purchase or view availability for this product.