Learn More
Sino Biological P53 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100035T3250
Produced in rabbits immunized with A synthetic phospho-peptide corresponding to residues surrounding Ser392 of human P53., and purified by antigen affinity chromatography.
Catalog No. 50-161-0323