Learn More
Sino Biological HER2 / ErbB2 / CD340 Antibody, Mouse MAb DFS Item
Manufacturer:  Sino Biological 10004MM01100
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human ErbB2 / HER2 extracellular domain (rhErbB2; Catalog#10004-H08H; NP_004439.2; Met 1-Thr 652). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-161-0324