Learn More
Sino Biological HER2 / ErbB2 Antibody, Rabbit PAb DFS Item
Manufacturer:  Sino Biological 10004RP01100
Produced in rabbits immunized with purified, recombinant Human HER2 / ErbB2 (rh HER2 / ErbB2; Catalog#10004-H20B1; NP_004439.2; Lys676-Val1255). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-161-0360