Learn More
Sino Biological HER2 / ErbB2 / CD340 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 10004T60100
Produced in rabbits immunized with purified, recombinant Human HER2 / ErbB2 (rh HER2 / ErbB2; Catalog#10004-H20B1; NP_004439.2; Lys676-Val1255). HER2 / ErbB2 specific IgG was purified by Human HER2 / ErbB2 affinity chromatography.
Catalog No. 50-161-0371