Learn More
Sino Biological CD112 / Nectin-2 / PVRL2 Antibody, Rabbit Mab DFS Item
Manufacturer:  Sino Biological 10005R0051
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD112 extracellular domain (rh CD112; Catalog#10005-H08H; Met 1-Leu 360; NP_002847.1).
Catalog No. 50-161-0377