Learn More
Sino Biological CD112 / Nectin-2 / PVRL2 Antibody, Rabbit PAb DFS Item
Manufacturer:  Sino Biological 10005RP03200
Produced in rabbits immunized with purified, recombinant Human CD112 / Nectin2 extracellular domain (rhCD112; Catalog#10005-H08H; Met 1-Leu 360; NP_002847.1). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-161-0385