Learn More
Sino Biological Human G-CSF / CSF3 Protein (isoform b) DFS Item
Manufacturer:  Sino Biological 10007HNCE5
A DNA sequence encoding the human GCSF isoform b (NP_757373.1) (Ala30-Pro204) was expressed.
Catalog No. 50-161-3767