Learn More
Sino Biological G-CSF / CSF3 Antibody, Mouse MAb DFS Item
Manufacturer:  Sino Biological 10007MM0750
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human G-CSF (rh G-CSF; Catalog#10007-HNCE; NP_757373.1; Ala 30-Pro 204). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-161-0392