Learn More
Sino Biological G-CSF/CSF3 Antibody, Rabbit MAb DFS Item
Manufacturer:  Sino Biological 10007R2431
This antibody was obtained from a rabbit immunized with purified, recombinant Human G-CSF/CSF3 (rh G-CSF/CSF3; Catalog#10007-HNCE; NP_757373.1; Ala30-Pro204).
Catalog No. 50-161-0398