Learn More
Sino Biological VEGF121/VEGFA Antibody, Rabbit MAb DFS Item
Manufacturer:  Sino Biological 10008R1451
This antibody was obtained from a rabbit immunized with purified, recombinant Human VEGF121/VEGFA (rh VEGF121/VEGFA; Catalog#10008-HNAH; P15692-9; Met1-Arg147).
Catalog No. 50-161-0418