Learn More
Sino Biological VEGF121/VEGFA Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 10008T26200
Produced in rabbits immunized with purified, recombinant Human VEGF121/VEGFA (rh VEGF121/VEGFA; Catalog#10008-HNAB; P15692-9; Met1-Arg147). VEGF121/VEGFA specific IgG was purified by Human VEGF/VEGFA affinity chromatography.
Catalog No. 50-161-0431