Learn More
Sino Biological Endoglin / CD105 / ENG Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 100088RP02100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human CD105, and purified by antigen affinity chromatography.
Catalog No. 50-161-0412