Learn More
Sino Biological VEGF183/VEGFA Antibody, Rabbit PAb DFS Item
Manufacturer:  Sino Biological 10009RP01400
Produced in rabbits immunized with purified, recombinant Human VEGF183/VEGFA (rh VEGF183/VEGFA; Catalog#10009-HNAB; P15692-3; Met1-Arg209). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-161-0440