Learn More
Sino Biological VEGF183/VEGFA Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 10009RP02100
Produced in rabbits immunized with purified, recombinant Human VEGF183/VEGFA (rh VEGF183/VEGFA; Catalog#10009-HNAB; P15692-3; Met 1-Arg 209). VEGF183/VEGFA specific IgG was purified by Human VEGF183/VEGFA affinity chromatography.
Catalog No. 50-161-0441