Learn More
Sino Biological EPHB2 antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100091T08100
Produced in rabbits immunized with A synthetic peptide corresponding to the N-terminus of the human EPHB2., and purified by antigen affinity chromatography.
Catalog No. 50-161-0433