Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wheaton™ Wheaton™ 1000mL Media Bottle, Clear Type I

Manufactured from borosilicate glass which provides the best chemical resistance. Wheaton 1000mL Media Bottle, Clear Type I are available with black phenolic Polyethylene or Rubber lined caps.

Manufacturer:  Wheaton™ W219420

 View more versions of this product

Catalog No. 02-912-651


  Check Availability
Add to cart

Description

Description

  • Clear borosilicate glass bottles, non-graduated
  • Special pour lip facilitates pouring
  • Packaged in partitioned carton to help prevent scratch marks on bottles
  • Hazardous communication label included for each case for content ID and safety notification
  • Tamper evident seal keeps lidded bottles sealed to protect contents from accidental misuse
Specifications

Specifications

1000mL
Pour lip
Partitioned case
Media Bottle
Black
103mm
Screw
Clear
TYPE 1 BOROSILICATE
12 EA
No
38-430
231mm
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit