Learn More
Tecan Disposable Tips, 1000µL, Conductive, Non-filtered, Non-sterile, Tecan Pure Purity Level, 2304 tips/case
Supplier: Tecan 30000630
Genuine Tecan® 1000 µL disposable automation tips for Freedom EVO® Liquid Handling Arm™ (LiHa) or Fluent® Flexible Channel Arm™ (FCA). Hanging tip format, non-filtered, conductive. Tecan PURE purity level - tested and certified to be free of human DNA, Rnase, Dnase and PCR inhibitors. 24 x 96-tip hanging racks, 2,304 tips/sales unit. LiHa Disposable Tips have been carefully designed and validated to work specifically with Liquid Handling Arms™ on all Tecan platforms. They are produced to stringent standards of quality and reproducibility, ensuring reliable liquid handling performance. Manufacturing is fully automated in a controlled environment, using state-of-the-art production processes, integrated quality control procedures and premium raw materials. During production, each tip undergoes in-line optical inspection using a CCD camera and image recognition software, checking all functional parts to ensure consistent pipetting accuracy and precision.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.