Learn More
Sino Biological Sox2 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100106RP02200
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human Sox2, and purified by antigen affinity chromatography.
Catalog No. 50-161-0454