Learn More
Sino Biological Beta-Tubulin Loading Control Antibody, Mouse MAb DFS Item
Manufacturer:  Sino Biological 100109MM05T50
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with a synthetic peptide corresponding to the N-terminus of the human beta-tubulin. The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-161-0457