Learn More
Sino Biological Neuropilin-1 / NRP1 / CD304 Antibody, Rabbit PAb DFS Item
Manufacturer:  Sino Biological 10011RP01100
Produced in rabbits immunized with purified, recombinant Human Neuropilin-1 / NRP1 / CD304 (rh NRP1; Catalog#10011-H08H; NP_001019799.1; Met1-Lys644). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-161-0470