Learn More
Sino Biological Human VEGFR2 / Flk-1 / CD309 / KDR Protein (Fc Tag) DFS Item
Manufacturer:  Sino Biological 10012H02H50
A DNA sequence encoding the extracellular domain of human VEGFR2 (NP_002244.1) (Met 1-Glu 764) was fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-3789