Learn More
Sino Biological Human VEGFR2 / Flk-1 / CD309 / KDR Protein (Domain 2&3, His Tag) DFS Item
Manufacturer:  Sino Biological 10012H02H250
A DNA sequence encoding the human KDR (NP_002244.1) (Asp120-Lys327) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-3788