Learn More
Sino Biological Human VEGFR2 / Flk-1 / CD309 / KDR Protein (Domain 1&2&3, His Tag) DFS Item
Manufacturer:  Sino Biological 10012H08H150
A DNA sequence encoding the human KDR (NP_002244.1) (Met1-Lys327) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-3792