Learn More
Sino Biological VEGFR2 / KDR / Flk-1 / CD309 Antibody, Mouse MAb DFS Item
Manufacturer:  Sino Biological 10012MM06200
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human VEGFR2 / KDR / Flk-1 / CD309 (rh VEGFR2 / KDR / Flk-1 / CD309; Catalog#10012-H08H; NP_002244.1; Met1-Glu764). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-161-0523