Learn More
Sino Biological VEGFR2 / Flk-1 / CD309 / KDR Antibody, Rabbit PAb DFS Item
Manufacturer:  Sino Biological 10012RP01400
Produced in rabbits immunized with purified, recombinant Human VEGFR2 / Flk-1 / CD309 / KDR (rh KDR; Catalog#10012-H08H; NP_002244.1; Met1-Glu764). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-161-0544