Learn More
Sino Biological Bcl-2 Antibody, Rabbit MAb DFS Item
Manufacturer:  Sino Biological 100126R20450
This antibody was obtained from a rabbit immunized with a synthetic peptide corresponding to the N-terminus of the Human Bcl-2. The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-161-0485