Learn More
Sino Biological CD79a Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100128RP0250
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human CD79a, and purified by antigen affinity chromatography.
Catalog No. 50-161-0501