Learn More
Sino Biological KRT14 / Cytokeratin 14 Antibody, Rabbit MAb DFS Item
Manufacturer:  Sino Biological 100129R101100
This antibody was obtained from a rabbit immunized with purified, a synthetic peptide corresponding to the N-terminus of the human Cytokeratin 14.
Catalog No. 50-161-0508