Learn More
Sino Biological aFGF / FGF1 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 10013RP03200
Produced in rabbits immunized with purified, recombinant Human fibroblast growth factor acidic (rhFGF-acidic; Catalog#10013-HNAE; aa 16-155; AAA79245.1). aFGF / FGF1 specific IgG was purified by human aFGF affinity chromatography.
Catalog No. 50-161-0593