Learn More
Sino Biological Ki-67 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100130RP02200
Produced in rabbits immunized with purified, a synthetic peptide corresponding to the sequence of the Human Ki-67, and purified by antigen affinity chromatography.
Catalog No. 50-161-0567