Learn More
Sino Biological GFAP Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100139RP02200
Produced in rabbits immunized with purified, a synthetic peptide corresponding to the C-terminus of the Human GFAP, and purified by antigen affinity chromatography.
Catalog No. 50-161-0584