Learn More
Sino Biological Human bFGF / FGF2 Protein DFS Item
Manufacturer:  Sino Biological 10014HNAE500
A DNA sequence encoding the mature form of human bFGF (NP_001997.5) (Pro 143-Ser 288) was expressed, with an additional Met at the N-terminus.
Catalog No. 50-161-3810