Learn More
Sino Biological Human GM-CSF / CSF2 Protein DFS Item
Manufacturer:  Sino Biological 10015HNAY100
A DNA sequence encoding human GMCSF (NP_000749.2) (Ala18-Glu144) was expressed.
Catalog No. 50-161-3824