Learn More
Sino Biological GM-CSF / CSF2 Antibody, Rabbit PAb DFS Item
Manufacturer:  Sino Biological 10015RP01100
Produced in rabbits immunized with purified, recombinant Human GM-CSF / CSF2 (rh CSF2; Catalog#10015-H07H; NP_000749.2; Ala18-Glu144). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-161-0624