Learn More
Sino Biological GM-CSF / CSF2 Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 10015RP02200
Produced in rabbits immunized with purified, recombinant Human GM-CSF / CSF2 (rh GM-CSF / CSF2; Catalog#10015-H07H; NP_000749.2; Ala18-Glu144). GM-CSF / CSF2 specific IgG was purified by Human GM-CSF / CSF2 affinity chromatography.
Catalog No. 50-161-0628