Learn More
Sino Biological beta-Actin Loading Control Antibody, Mouse MAb DFS Item
Manufacturer:  Sino Biological 100166MM1050
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with the N-terminus of the Human beta Actin / ACTB. The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-161-0634