Learn More
Sino Biological beta Actin / ACTB Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100166T36100
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human beta Actin / ACTB, and purified by antigen affinity chromatography.
Catalog No. 50-161-0635