Learn More
Sino Biological MERS-CoV (NCoV / Novel coronavirus) Spike Protein S1 Antibody (N-terminal), Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100208RP02100
Produced in rabbits immunized with purified, a synthetic peptide corresponding to the N-terminus of the S1 subunit of MERS-CoV (NCoV / Novel coronavirus) spike glycoprotein (S protein), and purified by antigen affinity chromatography.
Catalog No. 50-161-0640