Learn More
Sino Biological Human IL12A / NKSF1 Protein (His Tag) DFS Item
Manufacturer:  Sino Biological 10021H08H500
A DNA sequence encoding the p35 subunit of human IL12, termed as IL12A (P29459) (Met 1-Ser 219) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-3836