Learn More
Sino Biological IL12A / IL-12A Antibody, Mouse MAb DFS Item
Manufacturer:  Sino Biological 10021MM05100
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human IL12A / IL-12A (rh IL12A / IL-12A; Catalog#10021-H08H; P29459; Met 1-Ser 219). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-161-0653