Learn More
Sino Biological IL12A / NKSF1 Antibody, Rabbit PAb DFS Item
Manufacturer:  Sino Biological 10021RP01100
Produced in rabbits immunized with purified, recombinant Human IL12A (rh IL12A; Catalog#10021-H08H; NP_000873.2; Met 1-Ser 219). Total IgG was purified by Protein A affinity chromatography .
Catalog No. 50-161-0658