Learn More
Sino Biological CD3E Antibody, Rabbit PAb, Antigen Affinity Purified DFS Item
Manufacturer:  Sino Biological 100216T32200
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the human CD3E., and purified by antigen affinity chromatography.
Catalog No. 50-161-0651