Learn More
Sino Biological 4EBP1 Antibody, Mouse MAb DFS Item
Manufacturer:  Sino Biological 10022MM01T50
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human 4EBP1 (rh 4EBP1; Catalog#10022-H07E; NP_004086.1; Ser2-Ile118). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-161-0666