Learn More
Sino Biological 4E-BP1 / EIF4EBP1 Antibody (PE), Mouse MAb DFS Item
Manufacturer:  Sino Biological 10022MM04P100
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human 4E-BP1 / EIF4EBP1 (rh 4E-BP1 / EIF4EBP1; Catalog#10022-H07E; NP_004086.1; Ser2-Ile118) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-161-0672