Learn More
Sino Biological 4E-BP1 / EIF4EBP1 Antibody, Rabbit MAb DFS Item
Manufacturer:  Sino Biological 10022R05850
This antibody was obtained from a rabbit immunized with purified, recombinant Human 4E-BP1 / EIF4EBP1 (rh 4E-BP1 / EIF4EBP1; Catalog#10022-H07E; NP_004086.1; Ser 2-Ile 118).
Catalog No. 50-161-0680